Learn More
Histone H3 (monomethyl K56) Rabbit, Clone: RM180, Abnova™
Rabbit monoclonal antibody raised against synthetic monomethyl peptide of human Histone H3 (monomethyl K56).
Supplier: Abnova Corporation MAB12798
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. This structure consists of approximately 146 bp of DNA wrapped around a nucleosome, an octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H3 family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6p22-p21.3. [provided by RefSeq]
Specifications
Histone H3 (monomethyl K56) | |
Monoclonal | |
Unconjugated | |
ELISA (0.2 ug/mL-1 ug/mL) Western Blot (1 ug/mL-2 ug/mL) The optimal working dilution should be determined by the end user. | |
H3/A/H3FA | |
Rabbit | |
Protein A purification | |
RUO | |
8350 | |
Human | |
Liquid |
ELISA, Western Blot | |
RM180 | |
Rabbit monoclonal antibody raised against synthetic monomethyl peptide of human Histone H3 (monomethyl K56). | |
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide) | |
HIST1H3A | |
A synthetic monomethyl peptide corresponding to residues surrounding K56 of Histone H3. | |
100 μg | |
Primary | |
This antibody reacts to Histone H3 monomethylated at Lysine 56. No cross reactivity with nonmodified Lysine 56 or other methylations in histone H3. | |
Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.