Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3, Monomethyl Lys9, p Thr6 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB211052G
Description
Histone H3 Polyclonal specifically detects Histone H3 in Human, Mouse, C. elegans samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
Histone H3 (p Thr6, Monomethyl Lys9) | |
Polyclonal | |
Western Blot, Immunocytochemistry/Immunofluorescence | |
Q71DI3 | |
H3C14 | |
This Histone H3 [p Thr6, Monomethyl Lys9] antibody was raised against synthetic monomethylated/phosphorylated peptide surrounding Lysine 9/ Threonine 6 of human Histone H3.2 [Swiss Prot Q71DI3]. | |
0.05 mL | |
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
126961 | |
Human, Mouse, C. elegans | |
IgG |
Western Blot, Immunocytochemistry, Immunofluorescence | |
DyLight 488 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, mouse, and S. cerevisiae. Predicted to react with many species including rat, chicken, Xenopus, Drosophila, and plant based on 100% sequence homology. | |
Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Product Content Correction