Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259175

Catalog No. NBP259175


Add to cart

Description

Description

Histone H3 ac Lys23 Polyclonal antibody specifically detects Histone H3 ac Lys23 in Human samples. It is validated for Western Blot, ELISA, Dot Blot.
Specifications

Specifications

Histone H3 (ac Lys23)
Unconjugated
Serum with 0.05% Sodium Azide
H3 histone, family 3A, H3.3AH3F3H3F3B, H3.3B, H3K23ac, histone H3.3, MGC87782, MGC87783
Rabbit
IgG
100 ul
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls
Primary
126961
Western Blot, ELISA, Dot Blot
Western Blot 1:250, ELISA 1:100 - 1:500, Dot Blot 1:1000
Antiserum
HIST2H3C
The exact immunogen is propietary information.
Whole Antiserum
RUO
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only