Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3 (trimethyl K36) Rabbit, Clone: RM155, Abnova™

Rabbit monoclonal antibody raised against synthetic trimethyl peptide of human histone H3 (trimethyl K36).

Manufacturer:  Abnova Corporation MAB12835

Catalog No. 89-947-508


Add to cart

Description

Description

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. This structure consists of approximately 146 bp of DNA wrapped around a nucleosome, an octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H3 family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6p22-p21.3. [provided by RefSeq]

Specifications

Specifications

Histone H3 (trimethyl K36)
RM155
Rabbit monoclonal antibody raised against synthetic trimethyl peptide of human histone H3 (trimethyl K36).
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
H3/A|H3FA
Rabbit
IgG
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
This antibody reacts to histone H3 trimethylated at Lysine 36. No cross reactivity with monomethylated Lysine 36 or dimethylated Lysine 36, or other methylations in histone H3.
ChIP assay, ELISA, Western Blot
Unconjugated
ChIP (2-10 ug/mL) ELISA (0.2-1 ug/mL) Western Blot (0.5-2 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
HIST1H3A
A synthetic trimethyl peptide corresponding to residues surrounding K36 of human histone H3.
Protein A affinity purification
RUO
Primary
8350
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit