Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3, Trimethyl Lys36 Antibody (RM491), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP326038AF350
Description
Histone H3 Monoclonal antibody specifically detects Histone H3 in Human, Vertebrate samples. It is validated for ELISA, ChIP assay (ChIP), Multiplex ImmunoassaySpecifications
Histone H3 | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human, Vertebrates | |
Purified |
ELISA, ChIP Assay, Multiplex | |
RM491 | |
ELISA, Chromatin Immunoprecipitation (ChIP), Multiplex Immunoassay | |
H3, H3 histone, family 3A, H3.3A, H3.3B, H3F2, H3F3, H3F3B, H3FM, HIST2H3C, histone H3.3, MGC87782, MGC87783 | |
A trimethyl-peptide corresponding to Histone H3 [Lys36]. (Uniprot # P84243) | |
0.1 mL | |
Chromatin Modifiers, Chromatin Research, Epigenetics, Histones and Modified Histones, Loading Controls | |
126961 | |
Store at 4C in the dark. | |
IgG |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction