Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H3ac (pan-acetyl) Polyclonal Antibody, Active Motif™

Rabbit Polyclonal Antibody
$145.44 - $588.98
Specifications
Antigen | Histone H3ac (pan-acetyl) |
---|---|
Concentration | Conc. Not Determined |
Applications | ChIP assay, ChIP-sequencing, Dot Blot, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-3908
![]() ![]() |
Active Motif
39140 |
10 μL |
Each for $145.44
|
|
50-199-3906
![]() ![]() |
Active Motif
39040 |
50 μL |
Each for $345.42
|
|
50-199-3907
![]() ![]() |
Active Motif
39139 |
100 μL |
Each for $588.98
|
|
Description
Histone H3 is one of the DNA-binding proteins found in the chromatin of all eukaryotic cells. H3 along with four core histone proteins binds to DNA forming the structure of the nucleosome. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Post translationally, histones are modified in a variety of ways to either directly change the chromatin structure or allow for the binding of specific transcription factors. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.Specifications
Histone H3ac (pan-acetyl) | |
ChIP assay, ChIP-sequencing, Dot Blot, Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
This Histone H3 acetyl antibody was raised against a peptide including acetyl-lysines contained in the N-terminal tail of histone H3. | |
Primary | |
Antibody |
Conc. Not Determined | |
Polyclonal | |
Liquid | |
RUO | |
whole serum with 30% glycerol and 0.035% sodium azide; pH | |
IgG | |
-20°C, Avoid Freeze/Thaw Cycles |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title