Learn More
Histone H3K23me3 Rabbit anti-Human, Unconjugated, Polyclonal, Active Motif™
Rabbit Polyclonal Antibody
Manufacturer:  Active MotifSupplier Diversity Partner 61499
Encompass
 View more versions of this product
Catalog No. 50-199-3453
Add to cart
Description
Description
Histone H3 is one of the DNA-binding proteins found in the chromatin of all eukaryotic cells. H3 along with four core histone proteins binds to DNA forming the structure of the nucleosome. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Post translationally, histones are modified in a variety of ways to either directly change the chromatin structure or allow for the binding of specific transcription factors. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.
Specifications
Specifications
Histone H3K23me3
Lot-Specific
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
Rabbit
IgG
100 μL
-20°C, Avoid Freeze/Thaw Cycles
Primary
Human
Dot Blot
Unconjugated
Liquid
This antibody was raised against a peptide including trimethyl lysine 23 of human Histone H3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal