Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3K23me3 Rabbit anti-Human, Unconjugated, Polyclonal, Active Motif™ Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

$135.30 - $534.00

Specifications

Monoclonal or Polyclonal Polyclonal
Primary or Secondary Primary
Antigen Histone H3K23me3
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3454 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
61500
10 μL
Each for $135.30
Add to cart
 
50-199-3453 Non-distribution product as customer accommodation.
View Documents
Active MotifSupplier Diversity Partner
61499
100 μL
Each for $534.00
Add to cart
 
Description

Description

Histone H3 is one of the DNA-binding proteins found in the chromatin of all eukaryotic cells. H3 along with four core histone proteins binds to DNA forming the structure of the nucleosome. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Post translationally, histones are modified in a variety of ways to either directly change the chromatin structure or allow for the binding of specific transcription factors. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.
Specifications

Specifications

Polyclonal
Primary
Histone H3K23me3
Unconjugated
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit