Learn More
Histone H3K27me2me3 Mouse anti-Human, Mouse, Clone: 7B11, Active Motif™
Mouse Monoclonal Antibody
Manufacturer:  Active MotifSupplier Diversity Partner 39535
Encompass
 View more versions of this product
Catalog No. 50-199-4038
Add to cart
Description
Description
Histone H3 is one of the DNA-binding proteins found in the chromatin of all eukaryotic cells. H3 along with four core histone proteins binds to DNA forming the structure of the nucleosome. Histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Post translationally, histones are modified in a variety of ways to either directly change the chromatin structure or allow for the binding of specific transcription factors. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of post-translational modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine.
Specifications
Specifications
Histone H3K27me2me3
ChIP assay, Western Blot
Conc. Not Determined
ascites with 30% glycerol and 0.035% sodium azide; pH
P84244
Mouse
IgG
RUO
Antibody
Monoclonal
Human, Mouse
H3F3A, H3F3B
7B11
Unconjugated
H3F3B
Liquid
This Histone H3 di/trimethyl Lys27 antibody was raised against a peptide including trimethyl-lysine 27 of histone H3.
100 μL
-20°C, Avoid Freeze/Thaw Cycles
Primary
15078, 15081