Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H3Kme3Q5Ser Rabbit anti-Human,Mouse, KHL, Polyclonal, MilliporeSigma™

Rabbit Polyclonal Antibody

$153.50 - $394.00

Specifications

Antigen Histone H3Kme3Q5Ser
Applications ChIP assay, Dot Blot, Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ABE258010MI
View Documents
MilliporeSigma
ABE2580100UL
100 μL
Each for $394.00
Add to cart
 
ABE258025MI
View Documents
MilliporeSigma
ABE258025UL
25 μL
Each for $153.50
Add to cart
 
Description

Description

Histone H3Kme3Q5Ser Polyclonal antibody specifically detects Histone H3Kme3Q5Ser in Human,Mouse samples. It is validated for ChIP-seq, Dot Blot, Western Blotting.
Specifications

Specifications

Histone H3Kme3Q5Ser
KHL
Rabbit
H3KmeQ5Ser
KLH-conjugated linear peptide corresponding to 10 amino acids from the N-terminal region of human Histone H3 trimethylated on lysine 4 and serotonylated on glutamine 5.
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
ChIP assay, Dot Blot, Western Blot
Purified
Human, Mouse
HIST3H3, H3FT
Polyclonal
Affinity Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit