Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H4(acetyl K16) Rabbit, Clone: RM204, Abnova™

Rabbit monoclonal antibody raised against synthetic acetyl peptide of human Histone H4 (acetyl K16).

Manufacturer:  Abnova Corporation MAB12787

Catalog No. 89-947-512


Add to cart

Description

Description

Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Nucleosomes consist of approximately 146 bp of DNA wrapped around a histone octamer composed of pairs of each of the four core histones (H2A, H2B, H3, and H4). The chromatin fiber is further compacted through the interaction of a linker histone, H1, with the DNA between the nucleosomes to form higher order chromatin structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails; instead, they contain a palindromic termination element. [provided by RefSeq]

Specifications

Specifications

Histone H4(acetyl K16)
RM204
Rabbit monoclonal antibody raised against synthetic acetyl peptide of human Histone H4 (acetyl K16).
In PBS (50% glycerol, 1% BSA, 0.09% sodium azide)
H4/p|MGC24116
Rabbit
IgG
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
This antibody reacts to Histone H4 acetylated at Lysine 16. No cross reactivity with other acetylated Lysines in Histone H4.
ELISA, Western Blot
Unconjugated
ELISA (0.5 ug/mL-1 ug/mL) Western Blot (0.5 ug/mL-2 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
HIST4H4
A synthetic acetyl peptide corresponding to residues surrounding K16 of Histone H4.
Protein A purification
RUO
Primary
121504
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit