Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H4 Mouse anti-Human, Alexa Fluor™ 750, Clone: 282, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP242860AF750

Catalog No. NP242860A70


Add to cart

Description

Description

Histone H4 Monoclonal antibody specifically detects Histone H4 in Human samples. It is validated for Western Blot,Immunocytochemistry,Immunofluorescence.
Specifications

Specifications

Histone H4
282
Western Blot, Immunocytochemistry/Immunofluorescence
H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116
Mouse
IgG2b
Store at 4°C in the dark.
Monoclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
HIST4H4
The exact sequence is proprietary.
0.1 mL
Primary
121504
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit