Learn More
Histone H4 Mouse anti-Human, Clone: C.15200218, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259241
Catalog No. NBP259241
Add to cart
Description
Description
Histone H4 ac Lys12 Monoclonal antibody specifically detects Histone H4 ac Lys12 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation.
Specifications
Specifications
Histone H4 (ac Lys12)
C.15200218
Western Blot 1:500, Chromatin Immunoprecipitation 1-2 ug
Purified
HIST4H4
The exact immunogen is propietary information.
Protein A purified
RUO
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Monoclonal
Human
Western Blot, ChIP assay
Unconjugated
PBS with 0.05% Sodium Azide
H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, H4K12ac, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116
Mouse
IgG1
50 ug
Chromatin Modifiers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Histones and Modified Histones
Primary
121504