Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone H4, ac Lys5, ac Lys8, ac Lys12 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP259252
Description
Histone H4 Polyclonal specifically detects Histone H4 in Human, Mouse samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Immunocytochemistry/Immunofluorescence, Dot Blot, Chromatin Immunoprecipitation Sequencing.Specifications
Histone H4 (ac Lys5, ac Lys8, ac Lys12) | |
Polyclonal | |
Western Blot 1:1000, ELISA 1:1000, Immunocytochemistry/Immunofluorescence 1:500, Dot Blot 1:20000, Chromatin Immunoprecipitation (ChIP) 0.5-1 ug/IP, Chromatin Immunoprecipitation Sequencing | |
H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, H4K12ac, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116 | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse | |
IgG |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Dot Blot | |
Unconjugated | |
PBS with 0.05% Sodium Azide and 0.05% ProClin 300 | |
HIST4H4 | |
The exact immunogen is propietary information. | |
50 μg | |
Chromatin Modifiers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Histones and Modified Histones | |
121504 | |
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction