Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Histone H4 Rabbit anti-Human,Mouse,Xenopus, KHL, Clone: 5J18, MilliporeSigma™

Rabbit Monoclonal Antibody

Specifications

Antigen Histone H4
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Conjugate KHL
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ZRB100025MI
View Documents
MilliporeSigma
ZRB100025UL
25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
ZRB10004XMI
View Documents
MilliporeSigma
ZRB10004X25UL
4 x 25 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description

Description

Histone H4 Monoclonal antibody specifically detects Histone H4 in Human,Mouse,Xenopus samples. It is validated for Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting.
Specifications

Specifications

Histone H4
KHL
Rabbit
H4 histone family member A;Histone 1 H4a
KLH-conjugated linear peptide corresponding to 10 amino acids from the N-terminal region of human Histone H4.
Primary
Recommend storage of lyophilized product at 2 to 8°C. Avoid repeated freeze-thaws.
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Purified
Human, Mouse, Xenopus
HIST1H4A, H4/A, H4FA
Monoclonal
Protein A Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit