Learn More
Histone H4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP259247
Catalog No. NBP259247
Add to cart
Description
Description
Histone H4 Methyl Lys20 Polyclonal antibody specifically detects Histone H4 Methyl Lys20 in Human samples. It is validated for Western Blot, Chromatin Immunoprecipitation, ELISA, Dot Blot.
Specifications
Specifications
Histone H4 (Methyl Lys20)
Unconjugated
Serum with 0.05% Sodium Azide
H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/K, H4/M, H4/N, H4/O, H4/p, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO, H4K20Me, HIST1H4A, HIST1H4B, HIST1H4C, HIST1H4D, HIST1H4E, HIST1H4F, HIST1H4H, HIST1H4I, HIST1H4J, HIST1H4K, HIST1H4L, HIST2H4, HIST2H4A, HIST2H4B, histone 4, H4, histone cluster 4, H4, histone H4, MGC24116
Rabbit
IgG
100 ul
Chromatin Modifiers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Epigenetics, HA Epitope Tags, Histones and Modified Histones
Primary
121504
ChIP assay, Dot Blot, ELISA, Western Blot
Western Blot 1:750, Chromatin Immunoprecipitation 5 - 10 uL, ELISA 1:300, Dot Blot 1:20000
Antiserum
HIST4H4
The exact immunogen is propietary information.
Whole Antiserum
RUO
Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Polyclonal
Human