Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Histone macroH2A1.2 Monoclonal Antibody (14G7-H9), Active Motif™
Mouse Monoclonal Antibody
$147.50 - $565.00
Specifications
Antigen | Histone macroH2A1.2 |
---|---|
Clone | 14G7-H9 |
Concentration | Conc. Not Determined |
Applications | Immunocytochemistry, Immunofluorescence, Western Blot |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-4161
|
Active Motif
61428 |
10 μL |
Each for $147.50
|
|
|||||
50-199-4160
|
Active Motif
61427 |
100 μL |
Each for $565.00
|
|
|||||
Description
Histones are basic nuclear proteins that are responsible for the nucleosome structure of the chromosomal fiber in eukaryotes. Two molecules of each of the four core histones (H2A, H2B, H3, and H4) form an octamer, around which approximately 146 bp of DNA is wrapped in repeating units, called nucleosomes. The linker histone, H1, interacts with linker DNA between nucleosomes and functions in the compaction of chromatin into higher order structures. This gene is intronless and encodes a member of the histone H4 family. Transcripts from this gene lack polyA tails but instead contain a palindromic termination element. This gene is found in the large histone gene cluster on chromosome 6.Specifications
Histone macroH2A1.2 | |
Conc. Not Determined | |
Monoclonal | |
Liquid | |
RUO | |
ascites with 30% glycerol and 0.035% sodium azide; pH | |
Core histone macro-H2A.1; H2A histone family member Y; H2A histone family, member Y; H2A.y; H2A/y; H2AF12M; H2afy; Histone H2A.y; Histone macroH2A1; histone macroH2A1.1; histone macroH2A1.2; MACROH2A1; MACROH2A1.1; macroH2A1.2; Medulloblastoma antigen MU-MB-50.205; mH2A1 | |
MACROH2A1 | |
IgG2b | |
-20°C, Avoid Freeze/Thaw Cycles | |
MACROH2A1 |
14G7-H9 | |
Immunocytochemistry, Immunofluorescence, Western Blot | |
Unconjugated | |
Mouse | |
Human | |
O75367 | |
9555 | |
Recombinant protein corresponding to amino acids 198-228 of human Histone macroH2A1.2. | |
Primary | |
Antibody |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title