Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HistoneH2a Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP262793

Catalog No. NBP262793


Add to cart

Description

Description

HistoneH2a Polyclonal antibody specifically detects HistoneH2a in Human samples. It is validated for Western Blot, ELISA, Dot Blot, Chromatin Immunoprecipitation (ChIP).
Specifications

Specifications

HistoneH2a
Unconjugated
PBS with 0.05% Sodium Azide and 0.05% ProClin 300
H2A histone family, member Q, H2A/q, H2AFQH2A, H2A-GL101, histone 2, H2ac, histone cluster 2, H2ac, histone H2A type 2-C, Histone H2A/q, Histone H2A-GL101, histone IIa, MGC74460
Rabbit
IgG
50μg
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human
Dot Blot, ELISA, Immunoprecipitation, Western Blot
Western Blot 1:200, ELISA 1:200, Dot Blot 1:20000, Chromatin Immunoprecipitation (ChIP) 2 - 5μg
Affinity Purified
HIST2H2AC
Region of histone H2A containing the ubiquitinylated lysine 119 (H2AK119ub)
Affinity purified
Chromatin Research, Epigenetics, Histones and Modified Histones
Primary
8338
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit