Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Starplex™ Scientific Histoplex™ Tissue Cassettes with Micromesh Chamber

Innovative micromesh chamber securely retains fragments, leaving ample room for additional larger specimens

Manufacturer:  Starplex™ Scientific TCTAN

 View more versions of this product

Catalog No. 22-146-426

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »


Description

Description

  • Typical-looking tissue cassette designed with a corner section for use as a biopsy cassette
  • Fine micromesh chamber (0.23mm) is ideal for tissue fragments
  • Effective for embedding, processing, and storing standard tissue specimens and fragments in the same cassette
  • Durable acetyl polymer resists the chemical action of histological agents
  • Flow-through slots maximize fluid exchange
  • Textured 45° writing angle, suitable for labeling devices
  • Clam-shell design snaps closed and reopens with ease
  • Reduce inventory without giving up quality
  • Available in an assortment of non-metallic colors
  • Conveniently packaged 500 cassettes per dispenser box, three boxes per case

Specifications

Specifications

Slots
Clam-Shell
Acetal Polymer
45°
Tan
Tissue/Biopsy Cassette
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit