Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
Thermo Scientific™ HistoScreen™ Cassettes in Tube Packs
Handle small biopsies and fragile specimens with Thermo Scientific™ HistoScreen™ Cassettes in Tube Packs.
Manufacturer:  Thermo Scientific™ C750PK
 View more versions of this product
Catalog No. C750PK
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Handle small biopsies and fragile specimens with Thermo Scientific™ HistoScreen™ Cassettes in Tube Packs. Designed for automatic cassette labelers, they are the ultimate tube-fed cassettes for specimen security.
  • Saves time, offers the greatest specimen security, and provides ease of use
  • Provides an inner chamber lined with a “see-through” mesh for easy identification of specimens
  • Eliminates the need for messy sponges or lens paper
  • Ensures your tissues are safely processed with cassettes which will not pop open during processing
Specifications
Specifications
HistoScreen™ Cassettes, Mesh Pore Insert
Pink