Learn More
Simport™ Scientific Histosette™ II Tissue Processing/Embedding Cassette
Designed to hold tissue specimens during the embedding process, as well as in a storage file. Histosette™ II Tissue Processing/Embedding Cassettes also keep specimens safely submerged in liquid.
Supplier: Simport™ Scientific M4927
Description
- Made of acetal
- Keeps specimens safely submerged in liquid
- Resistant to the chemical action of most histological solvents
- 45° angle anterior writing area
- One-piece integral lid eliminates need for separate steel lids
- Back mounted tab allows for an unobstructed anterior marking area
- Open and close as often as necessary - cassettes will always relock securely Suitable for automated printers
Order Info
Each case contains 3 dispenser boxes of 500 cassettes.
Certifications
CE Marked
Specifications
Yes | |
Peach | |
Acetal | |
Tissue Cassette |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For in vitro diagnostic (IVD) use only.