Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare HisTrap™ FF Columns: 5 x 5mL: PROMO

Optimize purifications of histidine-tagged proteins directly from homogenized, unclarified cells HisTrap™ FF Columns: 5 x 5mL are prepacked with precharged Ni Sepharose™ 6 Fast Flow media.

Manufacturer:  GE Healthcare 17525501PM

Catalog No. 45-000-326LR

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • Prepacked with precharged Ni Sepharose™ 6 Fast Flow media
  • Column material: polypropylene
  • Bed dimensions: 7 × 25mm
  • Protein binding capacity of ∽40mg/mL media with negligible nickel ion leakage
  • Compatible with wide range of reducing agents, detergents, denaturants, and other additives
  • Simple operation with syringe, perstaltic pump, or chromatographic system
  • Columns may be connected in series to increase capacity
Medium
  • Matrix: 6% cross-linked agarose
  • Average particle size: 90μm
  • pH stability working range: 3 to 12
Specifications

Specifications

5mL
7mm
Polypropylene
3 to 12
5 bar
4° to 30°C in 20% ethanol
0.2 in.
<4mL/min.
5 × 5mL
HisTrap Column
5 × 5mL
6% Cross-linked Agarose
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit