Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva (Formerly GE Healthcare Life Sciences) HiTrap™ Desalting Prepacked Columns PROMO

For rapid convenient desalting and buffer exchange. HiTrap™ Desalting Prepacked Columns are prepacked, ready to use column for group separation between high and low molecular weight substances.

Manufacturer:  Cytiva (Formerly GE Healthcare Life Sciences) 29450315

Catalog No. 45-000-511RR0

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • Prepacked with Sephadex™ G-25 Superfine media
  • Column material: polypropylene
  • Bed volume: 5 mL
  • Bed dimensions: 16 x 25 mm
  • Sample volume: 1.5 mL
  • Flow velocity: <15 mL/min.
  • Maximum pressure: 3 bar

Media
  • Matrix: cross-linked dextran
  • Partical size: >20 µm
  • pH stability working range: 2 to 13

Specifications

Specifications

1 x 5 mL
HiTrap Desalting, 1 x 5 mL
>20 µm (dry), 15 to 88 µm (wet)
For Rapid Convenient Desalting and Buffer Exchange
4°C to 30°C, 20% Ethanol
HiTrap Desalting are ready-to-use 5 mL columns prepacked with Sephadex G-25 Superfine resin for fast and convenient desalting and buffer exchange.
No
0.62 in.
16 mm
5 mL
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit