Learn More
Cytiva HiTrap™ MabSelect™ Prepacked Columns PROMO
Cytiva HiTrap™ MabSelect™ Prepacked Columns PROMO for small-scale MAb purification in a continuous bioprocess
Manufacturer:  Cytiva 29489308
 View more versions of this product
Catalog No. 45-001-464LR0
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
Prepacked HiTrap columns for process development, screening of purification conditions, and small-scale purification of Mabs. MabSelect media are suited for the high flowrates and pressures required when scaling up.
  • Oriented coupling of recombinant Protein A ligand to the agarose matrix via an engineered C-terminal cysteine increases IgG binding capacity over other protein A media
  • High capacity, low ligand leakage, and high-flow base matrix allow purification of Mabs at process scale
  • Fewer regulatory concerns due to the total absence of mammalian culture in the ligand production and purification
  • Designed for use with a syringe, a peristaltic pump, or a chromatography system
MabSelect
  • Allows at least five times higher flowrates than conventional cross-linked agarose of similar porosity
  • Recombinant Protein A ligand promotes specificity of binding to Fc region of IgG; permits one-step purification
MabSelect Xtra™
  • Same recombinant protein A ligand as MabSelect, but with a smaller particle size and greater porosity, for increased dynamic binding capacity
  • Allows binding from samples with high expression levels of the monoclonal antibody
  • Lowers overall production cost due to the possibility of processing concentrated feedstocks in fewer batches
MabSelect SuRe™ (Superior Resistance)
  • Akali-tolerant protein A-derived ligand allows the use of up to 0.5M NaOH for cleaning-in-place (CIP)
  • Permits significant cost savings and extension of cycles in regular production
Specifications
Specifications
1.6 cm
< 20 mL/min.
20% Ethanol
HiTrap MabSelect
Fast purification of monoclonal antibodies from large sample volumes, Protein Purification
0.62 in.
≈ 30mg Human lgG/mL at 2.4 min.
Columns are prepacked with MabSelect, a BioProcess Resin for capturing of mAbs from large sample volumes.
5 mL
5 Pack