Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare HiTrap™ Protein G HP Prepacked Columns: PROMO

Use For fast, easy separations of IgG from different species and subclasses. GE Healthcare Life Sciences™ HiTrap™ Protein G HP Prepacked Columns are prepacked with Protein G Sepharose™.

Manufacturer:  GE Healthcare 29048581PM

 View more versions of this product

Catalog No. 45-000-504LR

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

  • Prepacked with Protein G Sepharose High Performance
  • Binding capacity: 25mg human IgG/column
  • Maximum pressure: 3 bar
  • Column material: polypropylene
  • For use with syringe, peristalic pump, or chromatography system
  • 1 and 5mL columns

Media
  • Ligand: Recombinant protein G lacking albumin-binding region
  • Matrix: 6% cross-linked agarose
  • Average particle size: 34μm
  • pH stability working range: 3 to 9

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.

5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.

Specifications

Specifications

25mg Human IgG/column
0.7cm
Fast, easy separations of IgG from different species and subclasses.
4° to 30°C
1 Pack
0.27 in.
<4mL/min.
20% Ethanol
Bed Dimensions: 7 x 25mm; 1 x 1mL
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit