Learn More
Alfa Aesar™ HIV Envelope Protein (254-274)
For cellular and molecular biology applications
Manufacturer:  Alfa Aesar™ J66539MCR
 View more versions of this product
Catalog No. AAJ66539MCR
Order in Bulk
Request bulk or custom formats
Add to cart
Specifications
Specifications
2208.56
1mg
Cys-Thr-His-Gly-Ile-Arg-Pro-Val-Val-Ser-Thr-Gln-Leu-Leu-Leu-Asn-Gly-Ser-Leu-Ala-Glu
-30° to -10°C
HPLC
Solid
RUO
HIV Envelope Protein (254-274)