Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
anti-HIV TAT specific factor 1, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Manufacturer: Novus Biologicals NBP118912
Description
HIV TAT specific factor 1 Polyclonal antibody specifically detects HIV TAT specific factor 1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
HIV TAT specific factor 1 | |
1.0 mg/mL | |
Western Blot 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:10-1:500 | |
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
cofactor required for Tat activation of HIV-1 transcription, dJ196E23.2, HIV TAT specific factor 1, HIV Tat-specific factor 1, HIV-1 Tat specific factor 1, TAT-SF1 | |
HTATSF1 | |
The immunogen recognized by this antibody maps to a region between residue 375 and 425 of human HIV-1 Tat specific factor 1 using the numbering given in entry NP_055315.2 (GeneID 27336). | |
Polyclonal | |
Immunogen affinity purified | |
RUO | |
Human |
Western Blot,Immunohistochemistry,Immunoprecipitation,Immunohistochemistry (Paraffin) | |
Unconjugated | |
Affinity Purified | |
NP_055315.2 | |
27336 | |
Rabbit | |
IgG | |
Primary | |
100 ul | |
Store at 4C. Do not freeze. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only