Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HIV TAT specific factor 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP30340920UL

 View more versions of this product

Catalog No. NB092542


Add to cart

Description

Description

HIV TAT specific factor 1 Polyclonal antibody specifically detects HIV TAT specific factor 1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence
Specifications

Specifications

HIV TAT specific factor 1
Unconjugated
Purified
Rabbit
IgG
20 μL
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
Immunofluorescence, Immunohistochemistry, Western Blot
PBS with 50% glycerol, pH7.3.
cofactor required for Tat activation of HIV-1 transcription, dJ196E23.2, HIV TAT specific factor 1, HIV Tat-specific factor 1, HIV-1 Tat specific factor 1, TAT-SF1
Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human HIV TAT specific factor 1 (NP_055315.2).
Affinity purified
RUO
Primary
27336
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit