Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HJURP Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A302822A

Description
The recommended shelf life for this product is 1 year from date of receipt.
Centromeric protein that plays a central role in the incorporation and maintece of histone H3-like variant CENPA at centromeres. Acts as a specific chaperone for CENPA and is required for the incorporation of newly synthesized CENPA molecules into nucleosomes at replicated centromeres. Prevents CENPA-H4 tetramerization and prevents premature DNA binding by the CENPA-H4 tetramer. Directly binds Holliday junctions.Specifications
HJURP | |
Polyclonal | |
Unconjugated | |
HJURP | |
Holliday junction recognition protein, 14-3-3-associated AKT substrate, Fetal liver-expressing gene 1 protein, Up-regulated in lung cancer 9, HJURP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55355 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q8NCD3 | |
HJURP | |
Between 650 and 700 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title