Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HJURP Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 152831AP

Catalog No. 50-173-0119


Add to cart

Description

Description

Centromeric protein that plays a central role in the incorporation and maintece of histone H3-like variant CENPA at centromeres. Acts as a specific chaperone for CENPA and is required for the incorporation of newly synthesized CENPA molecules into nucleosomes at replicated centromeres. Prevents CENPA-H4 tetramerization and prevents premature DNA binding by the CENPA-H4 tetramer. Directly binds Holliday junctions.
Specifications

Specifications

HJURP
0.25 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q8NCD3
FAKTS, FLEG1, hFLEG1, HJURP, Up regulated in lung cancer 9, URLC9
Rabbit
IgG
150 μL
-20°C
Primary
55355
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
HJURP
Liquid
HJURP
HJURP Fusion Protein Ag7402
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit