Learn More
HK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP
Rabbit Polyclonal Antibody
Supplier:  Proteintech Group Inc 156561AP150UL
Encompass_Preferred
 View more versions of this product
Catalog No. 50-173-0381
Add to Cart
Description
Description
This gene encodes a member of the ARHGAP family of Rho/Rac/Cdc42-like GTPase activating proteins. The encoded protein interacts with the Ras-related protein Gem through its N-terminal domain. Separately, it interacts with RhoA through a RhoGAP domain, and stimulates RhoA-dependent GTPase activity. Alternatively spliced variants that encode different protein isoforms have been described but their full-length nature has not been determined.
Specifications
Specifications
HK1
Polyclonal
Unconjugated
HK1
Hk1
HK1 Fusion Protein Ag8008
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Western Blot
0.16 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P05708, P17710, P19367
Rabbit
Antigen Affinity Chromatography
RUO
15275, 25058, 3098
-20°C
Liquid
Product Suggestions
Product Suggestions