Learn More
Aviva Systems Biology Corp HK2 Antibody - N-terminal region (ARP54303_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP54303_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against HK2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human HK2. Alternate names: DKFZp686M1669, HKII, HXK2ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-61167