Tocris™ HKI 357

Dual irreversible inhibitor of ErbB2 and EGFR

Manufacturer: Tocris™ 3580/10

 View more versions of this product

Catalog No. 35-801-0R

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only


    Specifications

    Inhibitors HKI 357
    Purity >98
    Molecular Weight 574.05
    Quantity 10mg
    Product Type HKI 357

    Potent, irreversible inhibitor of ErbB2 (HER2) and EGFR (IC50 values are 33 and 34nM respectively). Suppresses ligand-induced EGFR autophosphorylation and cell proliferation in NCI-H1975 cells containing L858R and T790M mutations. Circumvents mechanisms of resistance to Iressa (Cat. No. 3000) in non-small-cell lung cancer cells.

    • Chemical Name: (2E)-N-[[4-[[(3-Chloro-4-[(3-fluorophenyl)methoxy]phenyl]amino]-3-cyano-7-ethoxy-6-quinolinyl]-4-(dimethylamino)-2-butenamide
    • Formula: C31H29ClFN5O3
    • CAS Number: 848133-17-5
    • Solubility: Soluble to 100mM in DMSO and to 25mM in ethanol
    • Purity: >98

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit