Learn More
Raidians Bouton Lens Cleaning Papers
Highly absorbent
Manufacturer: Raidians 1BKVIS433
Description
- Safe to use with safety glasses and optical lenses
- Scratch and lint free
- High wet strength
- Papers are perforated for easy removal from booklets
Bibulous paper
- Heavy-duty
- Designed for blotting applications
- Lightweight
- Designed for use with spray lens cleaners
Specifications
Crystal lens cleaning papers | |
Cleaning lenses |