Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA-A Mouse anti-Human, FITC, Clone: 1082C5 (also known as 108-2C5), BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 568028
Description
HLA-A Monoclonal antibody specifically detects HLA-A in Human samples. It is validated for Flow Cytometry
Specifications
HLA-A | |
Monoclonal | |
FITC | |
HLA-A; HLAA; MHC class I antigen HLA-A heavy chain | |
Human Peripheral Blood Lymphocytes | |
RUO | |
Human | |
Liquid |
Flow Cytometry | |
1082C5 (also known as 108-2C5) | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Mouse | |
100 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction