Learn More
MBL International Corporation HLA-A2 100 U DFS Item
Manufacturer:  MBL International Corporation K01863
HLA-A2 Monoclonal Antibody Size 100ug; Clone Number BB7.2 Host Species mouse Conjugate Application FCM, IPP, IHC
Catalog No. 50-482-33