Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA Class I Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP275928

Catalog No. NBP275928


Add to cart

Description

Description

HLA Class I Polyclonal antibody specifically detects HLA Class I in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

HLA Class I
1 mg/mL
Western Blot 1:500-1:2000, Flow Cytometry 1:50-1:200 , Immunohistochemistry 1:200-1:500, Immunohistochemistry-Paraffin 1:200-1:500
Affinity Purified
HLA-A
Recombinant protein
40 kDa
100 μL
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
TBS (pH7.4), 0.5% BSA, 50% Glycerol with 0.05% Sodium Azide
A-10 alpha chain, FLJ26655, HLA class I histocompatibility antigen, A-1 alpha chain, HLA class I histocompatibility antigen, A-28 alpha chain, HLA class I histocompatibility antigen, A-9 alpha chain, HLAA, major histocompatibility complex, class I, A, MHC class I antigen A&asterisk;1, MHC class I antigen A&asterisk;11, MHC class I antigen A&asterisk;80
Rabbit
IgG
Peptide affinity purified
Adaptive Immunity, Cell Biology, Immunology
Primary
3105.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit