Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DMA Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen HLA DMA
Dilution Immunohistochemistry -1:200-1:1000, Immunohistochemistry-Paraffin -1:200-1:1000
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Conjugate Unconjugated
Format Purified
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121470
View Documents
Novus Biologicals
NBP311381200UL
200 μg
Each for $320.71
Add to cart
 
NB121471
View Documents
Novus Biologicals
NBP31138150UL
50 μg
Each for $138.37
Add to cart
 
Description

Description

HLA DMA Polyclonal antibody specifically detects HLA DMA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications

Specifications

HLA DMA
Immunohistochemistry, Immunohistochemistry (Paraffin)
Purified
RUO
Human
class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM
E. coli-derived Human HLA DMA fragment
Polyclonal
Antigen and protein A Affinity-purified
Immunohistochemistry -1:200-1:1000, Immunohistochemistry-Paraffin -1:200-1:1000
Unconjugated
Rabbit
Asthma, Diabetes Research, Immunology, Lipid and Metabolism, Membrane Trafficking and Chaperones, Neuroendocrinology
PBS, pH7.0
3108
IgG
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit