Learn More
HLA-DPB1 293T Cell Overexpression Lysate (Denatured), Abnova
Manufacturer: Abnova Corporation H00003115T01
Description
Western Blotting
Specifications
Western Blot | |
HLA-DPB1 293T Cell Transient Overexpression Lysate(Denatured) | |
29.2 | |
pCMV-HLA-DPB1 full-length | |
100 μL | |
Human |
293T | |
NM_002121 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-HLA-DPB1 antibody (H00003115-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) |