Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DQ/DR/DP Mouse anti-Human, Clone: rHLA-Pan/3475, Novus Biologicals™

Mouse Monoclonal Antibody

$159.82 - $374.34

Specifications

Antigen HLA DQ/DR/DP
Clone rHLA-Pan/3475
Concentration 0.2 mg/ml
Dilution Flow Cytometry 1-2 ug/million cells, ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunohistochemistry-Paraffin 1-2ug/ml
Applications Flow Cytometry, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB123089
View Documents
Novus Biologicals
NBP30780810UG
100 μg
Each for $374.34
Add to cart
 
NB123090
View Documents
Novus Biologicals
NBP30780820UG
20 μg
Each for $159.82
Add to cart
 
Description

Description

HLA DQ/DR/DP Monoclonal antibody specifically detects HLA DQ/DR/DP in Human samples. It is validated for Flow Cytometry, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications

Specifications

HLA DQ/DR/DP
0.2 mg/ml
Flow Cytometry, ELISA, Immunofluorescence, Immunohistochemistry (Paraffin)
Purified
RUO
Human
CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha
Non-T, non-B human acute lymphoblastic leukemia REH6 cells
Monoclonal
Protein A or G purified
rHLA-Pan/3475
Flow Cytometry 1-2 ug/million cells, ELISA 1-5 ug/ml, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunohistochemistry-Paraffin 1-2ug/ml
Unconjugated
Mouse
Adaptive Immunity, Asthma, Diabetes Research, Immunology
10 mM PBS with 0.05% BSA
3117
IgG1 Kappa
Primary
Store at 4°C.
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit