Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA DQ/DR/DP, Rabbit anti-Human, Clone: HLA-Pan/2967R, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP279709100UG

 View more versions of this product

Catalog No. NB010435


Add to cart

Description

Description

HLA DQ/DR/DP Monoclonal antibody specifically detects HLA DQ/DR/DP in Human Samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array.
Specifications

Specifications

HLA DQ/DR/DP
HLA-Pan/2967R
Unconjugated
10 mM PBS with 0.05% BSA
Purified
HLA-DQA1
Non-T, non-B human acute lymphoblastic leukemia REH6 cell line
Protein A or G purified
RUO
Store at 4°C.
Monoclonal
This antibody reacts with a common epitope of human major histocompatibility (MHC) class II antigens, HLA DQ/DR/DP.
ELISA, Flow Cytometry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA 2-5 ug/ml, Immunohistochemistry 1-2 ug/ml, Immunocytochemistry/Immunofluorescence 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
P04440, P01908, P01909, P01920, P01903, P01903
CD, CELIAC1DQ alpha 1 chain, DC-1 alpha chain, DQ-A1, FLJ27088, FLJ27328, GSE, HLA class II histocompatibility antigen, DQ(W3) alpha chain, HLA-DCA, HLA-DQA, leucocyte antigen DQA1, leukocyte antigen alpha chain, major histocompatibility complex, class II, DQ alpha 1, MGC149527, MHC class II antigen, MHC class II DQA1, MHC class II HLA-D alpha glycoprotein, MHC class II HLA-DQ-alpha-1, MHC class II surface glycoprotein, MHC HLA-DQ alpha
Rabbit
IgG
100 ug
Adaptive Immunity, Asthma, Diabetes Research, Immunology
Primary
3117
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit