Learn More
HLA-DQ Monoclonal Antibody (SK10), APC, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
$135.00 - $265.65
Specifications
Antigen | HLA-DQ |
---|---|
Clone | SK10 |
Concentration | 5 μL/Test |
Applications | Flow Cytometry |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-112-3235
|
Invitrogen
17988141 |
25 Tests |
Each for $135.00
|
|
|||||
50-113-0696
|
Invitrogen
17988142 |
100 Tests |
Each
for $265.65
Save 7% Reg : |
|
|||||
Description
Description: This SK10 monoclonal antibody reacts with the human MHC Class II HLA-DQ molecule. This cell surface receptor is a heterodimeric protein composed of an alpha (HLA-DQA1) and beta (HLA-DQB1) subunit. HLA-DQ is expressed on antigen presenting cells such as B cells, monocytes, macrophages, dendritic cells, and activated T cells. HLA-DQ plays a role in presenting peptide antigens to CD4+ T lymphocytes. Reports indicate that certain alleles of HLA-DQ are associated with celiac disease and type I diabetes. Applications Reported: This SK10 antibody has been reported for use in flow cytometric analysis. Applications Tested: This SK10 antibody has been pre-titrated and tested by flow cytometric analysis of normal human peripheral blood cells. This can be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Excitation: 633-647 nm; Emission: 660 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.
HLA-DQ Monoclonal antibody specifically detects HLA-DQ in Human samples. It is validated for Flow Cytometry.Specifications
HLA-DQ | |
5 μL/Test | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.2 | |
BoLA-DQA; BOLA-DQA1; BOLA-DQA2; BOLA-DQA2 protein; BOLA-DQA3; BOLA-DQA5; BOLA-DQB; CELIAC1; HLA class II antigen; HLA class II histocompatibility antigen, DQ beta 1 chain; HLA-DQB; HLA-DQB1; HLA-DRB1; IDDM1; lymphocyte antigen; major histocompatability complex class II; major histocompatibility complex, class II, DQ alpha 1 precursor; major histocompatibility complex, class II, DQ alpha 2; major histocompatibility complex, class II, DQ alpha 5; major histocompatibility complex, class II, DQ alpha, type 1; major histocompatibility complex, class II, DQ alpha, type 3; major histocompatibility complex, class II, DQ beta; major histocompatibility complex, class II, DQ beta 1; MHC class II antigen; MHC class II antigen DQB1; MHC class II antigen HLA-DQ-beta-1; MHC class II antigen precursor; MHC class II DQ beta chain; MHC class II DQA; MHC class II DQA precursor; MHC class II DQA1 precursor; MHC class II DQA2 (BoLA-DQA2); MHC class II DQA2 precursor; MHC class II DQB precursor; MHC class I | |
HLA-DQB1 | |
Primary | |
4° C, store in dark, DO NOT FREEZE! | |
HLA-DQB1 |
SK10 | |
Flow Cytometry | |
APC | |
Mouse | |
Human | |
P01920 | |
3119 | |
IgG1 κ | |
Affinity chromatography | |
Antibody |
For Research Use Only.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.