Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA-DQw1 Monoclonal Antibody (Genox 3.53), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Thermo Scientific MA146297
Description
Human Leukocyte Antigens are highly polymorphic proteins that are involved in the presentation of antigens to the T-cell receptor. There are two classes of HLA antigens, class I (HLA A, HLA B and HLA C) and class II (HLA D). This antibody can be used for HLA typing.Specifications
HLA-DQw1 | |
Monoclonal | |
0.7 mg/mL | |
HLA-DQB1 | |
HLA-DQB1 | |
Preparation of human Bristol 8 glycoprotein from the Bristol 8 B lymphoblastoid cell line. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Frozen) | |
Genox 3.53 | |
Unconjugated | |
P01920 | |
Mouse | |
Protein G | |
RUO | |
3119 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title