Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-ABC Mouse anti-Human, FITC, Clone: W6/32, Invitrogen™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen AHU0228

Catalog No. 50-113-7400


Add to cart

Description

Description

HLA-A belongs to the HLA class I heavy chain paralogues. This class I molecule is a heterodimer consisting of a heavy chain and a light chain (beta-2 microglobulin). The heavy chain is anchored in the membrane. Class I molecules play a central role in the immune system by presenting peptides derived from the endoplasmic reticulum lumen. They are expressed in nearly all cells. The heavy chain is approximately 45 kDa and its gene contains 8 exons. Exon 1 encodes the leader peptide, exons 2 and 3 encode the alpha1 and alpha2 domains, which both bind the peptide, exon 4 encodes the alpha3 domain, exon 5 encodes the transmembrane region, and exons 6 and 7 encode the cytoplasmic tail. Polymorphisms within exon 2 and exon 3 are responsible for the peptide binding specificity of each class one molecule. Typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. Hundreds of HLA-A alleles have been described.
Specifications

Specifications

HLA-ABC, HLA-DR (Class I)
Monoclonal
1 mg/mL
PBS with 5mg/mL BSA and 0.09% sodium azide; pH 7.2
P05534
HLA class I histocompatibility antigen, A-1 alpha chain, MHC class I antigen HLA-A heavy chain, MHC leukocyte antigen, antigen presenting molecule, leucocyte antigen A, leucocyte antigen class I, leukocyte antigen class I-A, MHC1, MHC 1
Mouse
IgG2a
100 tests
4° C, do not freeze
Primary
3105
Flow Cytometry
W6/32
FITC
HLA-A
Liquid
HLA-A
Cell membranes of human tonsil lymphocytes
Protein A
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit