Learn More
HLA-DR Mouse anti-Human, Pacific Orange™, Clone: TU36, Invitrogen™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  Invitrogen MHLDR30
Catalog No. 50-112-1484
Add to cart
Description
Description
Our Pacific Orange™ dye is optimally excited by the violet laser, and it is recommended that a 575, 585 or 600 nm band pass filter be used for optimal detection. Pacific Blue™ and Pacific Orange™ dye conjugates can be simultaneously excited at 405 nm and emit at 455 nm and 551 nm, respectively, facilitating two-color analysis.
HLA-DR, like other MHC class II molecules, is a transmembrane glycoprotein composed of a 36 kDa alpha chain (DRA) and 27 kDa beta chain (DRB). The alpha chain gene contains 5 exons. Exon 1 encodes the leader peptide, exons 2 and 3 encode the two extracellular domains, and exon 4 encodes the transmembrane domain and the cytoplasmic tail. DRA does not have polymorphisms in the peptide binding part and acts as the sole alpha chain for DRB1, DRB3, DRB4 and DRB5. Within the DR molecule the beta chain contains all the polymorphisms specifying the peptide binding specificities. Hundreds of DRB1 alleles have been described and typing for these polymorphisms is routinely done for bone marrow and kidney transplantation. HLA-DR is expressed primarily on antigen presenting cells such as B lymphocytes, monocytes, macrophages, thymic epithelial cells and activated T lymphocytes. Three loci, DR, DQ and DP, encode the major expressed products of the human class II region. The human MHC class II molecules bind intracellularly processed peptides, present them to T-helper cells, and have a critical role in the initiation of the immune response.
Specifications
Specifications
HLA-DR
Monoclonal
Pacific Orange™
HLA-DRB1
Liquid
HLA-DRB1
Human HLA-DR
Purified
ASR
Antibody
Monoclonal
Human
Flow Cytometry
TU36
PBS with BSA, sucrose and 0.1% sodium azide
P04229
HLA-DRA1, MLRW, HLA class II histocompatibility antigen, DR alpha chain, MHC cell surface glycoprotein, MHC class II antigen DRA, histocompatibility antigen HLA-DR alpha
Mouse
IgG2b
0.5 mL
4° C, store in dark
Primary
3123