Learn More
HLA-DR Mouse anti-Human, Clone: LN-3, Abnova™
Mouse monoclonal antibody raised against human HLA-DR.
Manufacturer:  Abnova Corporation MAB12723
Catalog No. 89-934-269
Add to cart
Description
Description
Specifications
Specifications
HLA-DR
LN-3
Mouse monoclonal antibody raised against human HLA-DR.
In tissue culture supernatant (0.7% BSA, 0.09% sodium azide).
Mouse
IgG2b
RUO
Primary
Human
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
Liquid
Human HLA-DR
250 uL
Store at 4°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal