Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HLA DRA Mouse anti-Human, Clone: 1C11A5, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP261863
Description
HLA DRA Monoclonal antibody specifically detects HLA DRA in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.Specifications
HLA DRA | |
1C11A5 | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
FLJ51114, histocompatibility antigen HLA-DR alpha, HLA class II histocompatibility antigen, DR alpha chain, HLA-DRA1, major histocompatibility complex, class II, DR alpha, MHC cell surface glycoprotein, MHC class II antigen DRA, MLRW | |
Mouse | |
IgG2b | |
Protein G purified | |
Adaptive Immunity, Cell Biology, Diabetes Research, Immunology | |
Primary | |
3122 |
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
1 mg/mL | |
Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:100-1:500 | |
Purified | |
HLA-DRA | |
Purified recombinant fragment of human HLA-DRA (AA: 26-254) expressed in E. Coli. | |
58.8 kDa | |
0.1 ml | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title