Learn More
Abnova™ HLA-DRB1 293T Cell Transient Overexpression Lysate (Denatured) (T04)
Human overexpression lysate
Manufacturer:  Abnova™ H00003123T04
Catalog No. 89-947-580
Add to cart
Specifications
Specifications
293T
BC008403
pCMV-HLA-DRB1 full-length
100 uL
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Western Blot
HLA-DRB1 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 30kDa
Transient overexpression cell lysate was tested with anti-HLA-DRB1 antibody (Mfr. No. H00003123-D01P) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Human