Learn More
HLA-E Mouse anti-Human, Alexa Fluor 647, Clone: 3D12 (also known as 3D12/HLA-E), BD Pharmingen™
Mouse Monoclonal Antibody
Supplier:  BD Biosciences 567418
 View more versions of this product
Catalog No. BDB567418
Add to Cart
This item is not returnable. View return policy
Description
Description
HLA-E Monoclonal antibody specifically detects HLA-E in Human samples. It is validated for Flow Cytometry
Specifications
Specifications
HLA-E
Monoclonal
Alexa Fluor 647
HLA class I histocompatibility antigen, alpha chain E; HLAE; MHC Class I Antigen E
Human HLA-E Recombinant Protein
RUO
Human
Liquid
Flow Cytometry
3D12 (also known as 3D12/HLA-E)
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse
100 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unreacted dye was removed.
IgG1 κ