Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HLA-E Mouse anti-Human, DyLight™ 650, Clone: 3D12HLA-E, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143124C

Catalog No. NBP143124C


Add to cart

Description

Description

HLA-E Monoclonal antibody specifically detects HLA-E in Human samples. It is validated for Western Blot,Flow Cytometry.
Specifications

Specifications

HLA-E
3D12HLA-E
Western Blot, Flow Cytometry
DKFZp686P19218, EA1.2, EA2.1, HLA class I histocompatibility antigen, alpha chain E, HLA class I histocompatibility antigen, E alpha chain, HLA-6.2, HLAE, lymphocyte antigen, major histocompatibility complex, class I, E, MHC, MHC class I antigen E, MHC HLA-E alpha-1, MHC HLA-E alpha-2.1, QA1
Mouse
IgG1 Kappa
Store at 4°C in the dark.
Monoclonal
Human
Flow Cytometry, Western Blot
DyLight™ 650
50 mM sodium borate with 0.05% sodium azide
HLA-E
The immunogen for this antibody was HLAE.
0.1 mL
Primary
3133
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit